Anti-ZNF185 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: United States Biological
USBI044140-FITCEA 852 EUR
USBI044140-FITC
Anti-ZNF185 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ZNF185 (zinc finger protein 185 (LIM domain))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR