Anti-MORN5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Supplier: US Biological
USBI038546-APCEA 852 EUR
USBI038546-APC
Anti-MORN5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: MORN5 (MORN repeat containing 5)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse