Anti-RASSF6 Goat Polyclonal Antibody

Supplier: Everest Biotechnology
EVBIEB08394EA 325.5 EUR
EVBIEB08394
Anti-RASSF6 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RASSF6 (Ras association (RalGDS/AF-6) domain family member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR