PerfectBlue™ Semi-Dry Electroblotters are designed for clean, rapid and effective transfer of proteins and nucleic acids from polyacrylamide or agarose gels to membranes. Suitable for use with Western, Southern and Northern blotting the Semi-Dry Blotters reduce many lengthy procedures down to around an hour.
Blotting chamber
Highly durable Plexiglass construction
Totally removable cover which attaches flat to reduce overall unit storage size
Oversized, easy to handle screws for fumble free and safe cover adjustment
High-quality gold plated plug contacts for optimal electrical contact
Electrodes
Solid electrode plates for an even transfer over the entire gel surface
High-grade stainless steel cathode and platinum-coated titanium anode for maximum transfer efficiencies
Sturdy cover for enhanced user safety
Design of lid prevents accidental wrong orientation of electrical field