Bottles, vials and tube racks, SP Bel-Art
Supplier: Bel-Art
BELAH189210000EA 57.9 EUR
BELAH189210000
Bottles, vials and tube racks, SP Bel-Art
Racks Bottle Racks
PP.
  • Stable, heavy duty, twelve wells with four front-open
  • Dimension: 267×102×53 mm
  • Good chemical and temperature resistance
Ideal for dropping bottles, vials, scintillation vials and other containers up to 28 - 33 mm diameter.