Block Heaters, Digital, HX-1 and HX-2
PEQL91-D1100EA 485 EUR
PEQL91-D1100 PEQL91-D1200
Block Heaters, Digital, HX-1 and HX-2
High-quality, thermostat-controlled block heaters for the incubation of a wide range of reaction tubes up to 150 °C.
  • Accurate microprocessor-controlled heating up to 150 °C
  • Target temperature or current temperature displayed on large, easy to read, LED display
  • Simple 'up/down' push button controls
  • Water resistant control panel and screen
  • RS232 interface transfers temperature data to a PC
  • Choice of single (HX-1) or double (HX-2) block models
Ordering information: Heating blocks are not included, please order these separately.