Hand cleanser, Stokosept® wash
Supplier: Deb Stoko
115-1481EA 0 EUR
EVON99063973 EVON99042969 EVON99042974 EVON99042976
Hand cleanser, Stokosept® wash
Liquid hand cleanser removes fats, protein and bacteria (incl. MRSA, VRE), viruses such as BVDV, RSV, HSV1, HIV, HBV.
  • Antibacterial (EN 1040, prEN 13727, EN 1499), fungistatic (EN 1275, EN 1650)
  • Skin friendly pH
  • Non soapy, alcohol-free, perfume-free, silicone-free
Active ingredient: 1,6 g Chlorhexidine digluconate/100 g product.