FoodChek™ MICT Reader
710-1595EA
10518.19
EUR
FCHKFCR-004
FoodChek™ MICT Reader
Rapid Food Pathogen Screening of Listeria, Salmonella and E.coli on food contact surfaces and food samples. The FoodChek™ MICT Reader uses magnetic nanotechnology via a compact, diagnostic reading device that reports test results from disposable assay cassettes preloaded with Actero™ enriched food samples. This desktop-based, user-friendly MICT Reader allows for automated tracking, recording and printing of quantitative test results without the need for a sophisticated technician or commercial lab.This allows for the fastest most accurate and easiest-to-use in-house pathogen testing systems in today's marketplace, is at least one order of magnitude more sensitive than PCR testing and a two order of magnitude higher than traditional (ELISA) tests and virtually eliminates human error. FoodChek™'s MICT Cleaning Cassettes (sold separately) are specifically designed for use with it's MICT Readers. The cleaning cassettes are single use only. FoodChek Systems Inc. recommends that the MICT reader should be cleaned after 20 tests or 1 month (whichever comes first) , in the event of an 'invalid result', when you are changing pathogen tests. ( E. coli to Listeria spp.). FoodChek™'s MICT Verification Cassette must be run at the start of the testing day and is specifically designed for use with the FoodChek™ MICT Reader. You can verify the FoodChek MICT Reader's accuracy and performance with the supplied verification cassette. This cassette contains three stable, calibrated test regions on one test strip. They are used to verify the instrument's internal calibration (Accuracy) and performance (Noise). The Verification Cassette is for multiple uses and only needs to be replaced when your VWR representative recommends to.
- Single-use FoodChek™ MICT Reader cleaning cassettes are sold separately
- ACTERO™ media used with Foodcheck MITC reader will make the sample enrichment phase up to 30% faster for E. coli O157 and Listeria and up to an amazing 70% faster for Salmonella
- Supplied with multiple-use, verification cassette
- Accurate and with high performance
Delivery information: 1 MICT reader, 1 user manual CD, 1 stylus, 1 verification cassette, 1 USB flash drive and 3 rolls of paper (enough for approx. 100 tests).
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Item(s) Added to Cart