Ultrasonic baths, DHA-1000
Supplier: Branson
BRAN000-914-606EA 4059.6 EUR
BRAN000-914-606
Ultrasonic baths, DHA-1000
The DHA-1000 ultrasonic bath is fully integrated, with ultrasonic generator and cleaning tank in a single enclosure. It is suitable for cleaning larger laboratory glassware/apparatus, fabricated metal sub-assemblies, printed circuit boards and injection molded plastic parts.
  • Stainless steel tank with rounded corners for easy cleaning
  • Analog controls mounted below a drip lip
  • On/off heat switch heats upto 60 °C