Chemistry kit, PYREX®
Supplier: Corning
PYREX®
CORN2705-14 CORN6412A-125 CORN6949K CORN2155-14 CORN9050-14
Chemistry kit, PYREX®
Microchemistry Kits
Chemistry kit is designed for most common organic chemistry experiments.
It comprises 15 components with 14/20 standard tapered joints including flasks, adapters, condensers. The components are held in foam within a polyethylene case.