Liquid nitrogen dewars with narrow neck for extended holding time, B Series
478-0598EA 1180 EUR
VWRI478-0598 VWRI478-0599 VWRI478-0600 VWRI478-0597
Liquid nitrogen dewars with narrow neck for extended holding time, B Series
Aluminium containers for storage in canisters. This generation of 'long holding time'' containers are designed for use in biology, research, pharmaceutical and medical fields, livestock breeding. They can be used for the storage of straws or cryovials. The tanks are manufactured in aluminium with very narrow fiberflass/epoxy neck.
  • Specially designed for livestock breeding (AI), Life Science and IVF
  • Canister storage system primally for the conservation of samples in straws (or cryo-vials)
  • For some models possibility to have a 2 straw levels canister, or long canes for cryovials
  • The high vacuum multidelayered super-insulation from the inner space limits nitrogen evaporation
  • The samples are placed in round stainless steel canisters (included) directly inside the vessel
  • For the user's safety and to avoid spillage, the containers have a lid latching device that can also be padlocked
Certifications: The units are made in France and manufactured according to the highest quality levels All units of the B 2000 series are CE-marked, and comply with the Medical Device Directive 93/42/CEE.