The CryoRoll 230 is built in stainless steel are designed for the road transport of cryogenic fluids in tough conditions. Self pressurised container for liquid nitrogen, oxygen and argon.
- For safety and easy transport, a handrail is also provided (handle + 4 castors - fork lift and truck fixation on demand)
This tank is equipped with a pressure regulator and a relief valve for a maximum work pressure till 3,9 bar. Filling/withdrawal and venting valves and a float level indicator are standard mounted on the vessels. For safety and easy transport, a handrail is also provided. Other capacities or horizontal versions upon request. When an accurate indication of the remaining liquid nitrogen is needed, a digital level indicator can be mounted instead of the float level gauge. It is battery operated and the analogue 4/20 mA signal can be remoted. A flexible or rigid elbow for LN₂ withdrawal can be mounted on the filling/withdrawal valve.
Certifications: The equipment is certified by the RID/ARD and complies with the Transportable Pressure Equipment Directive - TPED 10/35/UE.
In compliance with the European Directive TPED 99/36
Medical Device Certification 93/42 can be delivered if expressively required.