Low pressure LN₂ cylinder with roller base for transport, CryoRoll
478-0506EA 0 EUR
VWRI478-0506
Low pressure LN₂ cylinder with roller base for transport, CryoRoll
The CryoRoll 230 is built in stainless steel are designed for the road transport of cryogenic fluids in tough conditions. Self pressurised container for liquid nitrogen, oxygen and argon.
  • For safety and easy transport, a handrail is also provided (handle + 4 castors - fork lift and truck fixation on demand)
This tank is equipped with a pressure regulator and a relief valve for a maximum work pressure till 3,9 bar. Filling/withdrawal and venting valves and a float level indicator are standard mounted on the vessels. For safety and easy transport, a handrail is also provided. Other capacities or horizontal versions upon request. When an accurate indication of the remaining liquid nitrogen is needed, a digital level indicator can be mounted instead of the float level gauge. It is battery operated and the analogue 4/20 mA signal can be remoted. A flexible or rigid elbow for LN₂ withdrawal can be mounted on the filling/withdrawal valve.
Certifications: The equipment is certified by the RID/ARD and complies with the Transportable Pressure Equipment Directive - TPED 10/35/UE.
In compliance with the European Directive TPED 99/36
Medical Device Certification 93/42 can be delivered if expressively required.