Caspase uninduced lysate
Supplier: G-Biosciences
786-CULEA 260 EUR
GENO786-CUL
Caspase uninduced lysate
Caspase Uninduced Lysate is a simple tool for studying apoptosis and acts as an ideal negative control. Human Jurkat cells are used for preparing uninduced lysates.
Delivery information: It is supplied as a stabilised pellet along with CasPASE lysis buffer for pellet resuspension.