siRNA, iBONi® Box
Supplier: riboxx Life Sciences
XXXXD-00105-0005-R XXXXD-00105-0002-H XXXXD-00105-0002-M XXXXD-00105-0005-H XXXXD-00105-0002-R XXXXD-00105-0005-M
siRNA, iBONi® Box
Each iBONi® Box contains 4 pre-designed IBONI® siRNA (2 or 5 nmol), siRNA delivery reagent, siRNA controls and siRNA storage buffers.
Each iBONi® Box contains 4 pre-designed IBONI® siRNA (2 or 5 nmol), siRNA delivery reagent, siRNA controls and si RNA storage buffers.