miRNA inhibitors, CONTRAmiR®
Supplier: riboxx Life Sciences
XXXXM-00104-0010-X
miRNA inhibitors, CONTRAmiR®
CONTRAmiR® miRNA inhibitors are single-stranded nucleic acids from the class of antagonists of microRNA (15 to 25 nt). CONTRAmiR® effectively inhibits miRNA function with a subsequent change in phenotype of the cell and/or tissue. CONTRAmir® miRNA inhibitors display the complementary sequence to the mature miRNA. CONTRAmir® miRNA inhibitors in vivo display 6 to 9 phosphothioate groups in the backbone to enhance resistance to RNAses in body fluids.
  • For in vitro and in vivo applications
  • Pre-designed or custom miRNA inhibitors (custom modifications on request)
Ordering information: For the CONTRAmiR® inhibitor X-Mod the following chemical modifications can be ordered: 3'-Amino, 3'-Biotin, 3'-Chol, 3'-Peptide, 5'-Amino, 5'-6-FAM, 5'-Phosphate, 5'-Peptide, 5'-Thiol, internal: 2'-OMe, 2'-Fluoro, Inosine, PTO. Please contact your local VWR sales office for further information.