You Searched For: Skin Disinfectants
22  results were found
SearchResultCount:"22"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: GOJO Industries - Europe Ltd
Description: A no-rinse hygienic hand rub that feels great to use and is clinically proven to maintain skin health.
Supplier: GOJO Industries - Europe Ltd
Description: A no-rinse hygienic hand rub that feels great to use and is clinically proven to maintain skin health.
Supplier: GOJO Industries - Europe Ltd
Description: Spa-inspired foam hand wash offers luxurious lather and a pleasant botanical fragrance.
Safer Human and Environmental Health
Supplier: GOJO Industries - Europe Ltd
Description: A hygienic hand sanitising foam formulation, proven to help maintain skin health.
Catalog Number: (GOJO5179-03-EEU00)
Supplier: GOJO Industries - Europe Ltd
Description: Smooth foaming hand wash with broad spectrum germ killing power and moisturising properties. Hypoallergenic and mild to the skin for frequent usage.
UOM: 1 * 3 items
Supplier: Deb-STOKO
Description: This is a solvent-free workshop cream with plastic microbeads, effective against strong and adherent soiling such as greases, oils and fuels.
Supplier: Deb-STOKO
Description: This is a heavy duty workshop soap without microbeads, effective against strong and adherent soiling.
Supplier: Deb-STOKO
Description: Hypoallergenic, alcohol-based foam sanitiser for use in food industry. Compatible for use with latex and nitrile gloves.
MSDS Sale
Supplier: Diversey
Description: Disinfects hands in one easy step without the need for water. Non-classified highly efficacious alcohol-free foam hand rub with a refreshing fragrance...
Safer Human and Environmental Health
Sustainable Materials
Supplier: Diversey
Description: These SoftCareâ„¢ Des E disinfectant sprays, alcohol-based are effective against bacteria, virus and fungi. It is made up of 70% ethanol, which ensures ...
Catalog Number: (DIVY100860440)
Supplier: Diversey
Description: Soft Care Des E spray is a hand disinfectant based on alcohol (71,58%) and is applied without water. Soft Care Des E spray is suitable for use in all ...
UOM: 1 * 2 Can
Supplier: Hartmann Bode
Description: Sterillium® med is an ethanol based hand disinfectant designed for hygienic and surgical rub-in hand disinfection with good skin tolerability.
Catalog Number: (BODL9804100)
Supplier: Hartmann Bode
Description: The propanol based hand disinfectant is ideal for use in all areas of the health industry (operating theatres) and hygiene sensitive industries.
UOM: 1 * 100 mL
Supplier: BODE CHEMIE GMBH
Description: Sterillium® classic pure is particularly suitable for users with sensitive skin. The propanol based hand disinfectant is ideal for use in all areas of...
Supplier: SCHUELKE & MAYR GMBH
Description: Alcohol-based hand rub for hygienic and surgical hand disinfection with panthenol and vitamin E, without colour and perfume.
Catalog Number: (BODL9801440)
Supplier: Hartmann Bode
Description: Baktolan® Balm is a water-in-oil balm with high quality nourishing components that were particularly adapted to the skin’s pH value (5,5). This stabil...
UOM: 1 * 350 mL
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 22
no targeter for Bottom