You Searched For: Adhesion Testers
4  results were found
SearchResultCount:"4"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (SIMELD9200)
Supplier: Industrial Physics
Description: This instrument is an easy to use, lightweight hydraulic adhesion tester that applies an increasing level of pull off force to a dolly adhered to the ...
UOM: 1 * 1 items
Catalog Number: (SIMESP3000)
Supplier: Industrial Physics
Description: The TQC Cross Cut adhesion test kit (Master Paint Plate) is a stainless steel measuring tool that features: a 1, 1,5, 2 and 3 mm cross cut adhesion te...
UOM: 1 * 1 items
Catalog Number: (SIMESP3100)
Supplier: Industrial Physics
Description: This Adhesion Test Kit (HPK) is a method developped in the Netherlands by FOSAG and SKH to determine the adhesion of new and old paint layers on wood ...
UOM: 1 * 1 items
Catalog Number: (SIMELD9330)
Supplier: Industrial Physics
Description: This automatic adhesion tester measures the adhesion of coatings applied on metal, wood, concrete and similar substrates.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 4 of 4
no targeter for Bottom