Advanced gas filter system
Supplier: SGE (Trajan)
SGEA1034886EA 181 EUR
SGEA1034886 SGEA1034884 SGEA1034888
Advanced gas filter system
The advanced gas filter system has high capacity and efficiency levels for oxygen (750 cc, <1 ppb), water (12 g, <10 ppb) and hydrocarbons (8 g, <1 ppb). The recommended flow rate is 2 l/min and 200 psi maximum operating pressure.
The two part system includes a small stainless steel manifold and a high capacity filter cartridge. It can be attached to a laboratory wall or bench with the brackets included.
Each connection in the advanced gas filter system is made with two fluoro-elastomer seals. The polycarbonate shield that surrounds the glass indicator section of the filter is sealed, so the gas flow is secure even if the glass is broken.