Floor scales, MPC series for medical use
Supplier: Kern & Sohn
KERNMPC250K100MEA 306 EUR
KERNMPC250K100M
Floor scales, MPC series for medical use
Balances and Scales
Step-on personal floor scale with EC type and medical approval which can be used in medical diagnostics. It has second display on the rear of the balance.
  • Easy and hygenic cleaning
  • Secure, vibration free and non-slippery with height adjustable rubber feet
  • Automatic data hold function
  • Display can be mounted to the wall
Certifications: Please note: If a calibration certificate is required, this should be requested when ordering the balance, as it can not be issued retrospectively.