Gas purifiers, GateKeeper® Inert
Supplier: Entegris
568-0842EA 0 EUR
ENTE2500KFI4R2 ENTECE2500KFI4R ENTECE500KFHPI4R ENTECE2500KHI4R ENTECE2500KHI8R ENTECE300KFI4R ENTECE100KFI4R ENTECE10MHI8Y ENTECE500KFI4R ENTECE700KFI4R ENTECE70KFI4R ENTECE75KFI4R ENTECE30KFI4R ENTE35KFI4R2 ENTECE35KFI4R
Gas purifiers, GateKeeper® Inert
These purifiers are designed for use with N₂, He, Ne, Kr, Xe, Ar, halocarbons and light hydrocarbons. They remove O₂, H₂, CO, CO₂ and H₂O to sub-ppb (part-per-billion) levels; SO₂ to sub-ppt (part-per-trillion) levels.
  • Guards against bulk gas purifier breakthrough, impurity spikes from liquid storage tanks, cylinder changeouts and unexpected contaminants in the process
  • Does not release hydrocarbons that may contaminate the process
  • Uses ambient temperature purification technology – no power or heat required
  • No moving parts
CE certified to the Pressure Equipment Directive (PED) and manufactured in an ISO 9001:2000 facility.
Maximum inlet challenge of contaminant to maintain product efficiency: 10 ppm (parts-per-million)
Lifetime: One year at nominal flow rate with 1 ppm inlet challenge of moisture
Articles may not be available in all countries. Please contact your local VWR office.