Gas purifiers, GateKeeper® Optics
Supplier: Entegris
ENTECE2500KH08REA 0 EUR
ENTECE2500KH08R ENTECE300KF04R ENTECE10MH08Y ENTE2500KH08R2 ENTECE75KF04R ENTECE500KF04R ENTECE2500KF04R ENTECE70KF04R
Gas purifiers, GateKeeper® Optics
These purifiers are designed for use with CDA (clean dry air). They remove H₂O, volatile bases, volatile acids, condensable organics and refractory compounds to ppt (parts-per-trillion) levels.
  • Guards against bulk gas purifier breakthrough, impurity spikes from liquid storage tanks, cylinder changeouts and unexpected contaminants in the process
  • Does not release hydrocarbons that may contaminate the process
  • Uses ambient temperature purification technology – no power or heat required
  • No moving parts
CE certified to the Pressure Equipment Directive (PED) and manufactured in an ISO 9001:2000 facility.
Maximum inlet challenge of contaminant to maintain product efficiency: 10 ppm (parts-per-million)
Lifetime: One year at nominal flow rate with 1 ppm inlet challenge of moisture