Stainless steel floor scraper
Supplier: Micronova
129-0624EA 292 EUR
MICRQDFSRE-11
Stainless steel floor scraper
A heavy-duty, stainless steel floor scraper with a 279.4 mm (11") scraping surface. Ideal for cleaning dried residues from the floor and other flat surfaces in the cleanroom.
  • Heavy-duty floor scraping tool
  • Made from 304 electropolished stainless steel
  • Easy to grip red handle
  • Quick Disconnect handle attachment
This floor scraper is made from durable 304 electropolished stainless steel and can be attached to the SSU series Universal Handles for extended reach.
Ordering information: Handles must be ordered separately. For additional handle lengths, see SSU series Universal Mop Handles from Micronova.