Sponges, NovaLite™
Supplier: Micronova
NovaLite™
MICRSP2-46EA 16.3 EUR
MICRSP2-46
Sponges, NovaLite™
Wipes Sponges
These adaptable, hard-wearing sponges are ideal for applications in pharmaceutical, medical device and hi-tech cleanrooms. The exterior is made from Novalite material - a latex material bonded with cellulose. The sponge interior is cleanroom approved, reticulated urethane foam.
  • Absorbent and hard-wearing: affords greater friction resistance
  • Ideal for removing dust and fine powders
  • Resistant to isopropyl alcohol
  • Can be laundered and autoclaved