Sponges, NovaSponges™
Supplier: Micronova
NovaSponges™
MICRSP1-46EA 17.5 EUR
MICRSP1-46
Sponges, NovaSponges™
Wipes Sponges
These low linting, fast-drying sponges are made of a cleanroom approved reticulated urethane foam interior with a PVA cover. They are ideal for cleaning and wiping applications in microelectronics, semiconductor and aerospace environments.
  • Ideal for general cleaning, lapping and polishing
  • Can be laundered and re-used
  • Not recommended for use with isopropyl alcohol