You Searched For: Blood
4  results were found
SearchResultCount:"4"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Charles River Laboratories Cell Solutions
Description: Cord blood is known to be a rich source of hematopoietic stem cells, which can be used to research hematopoietic and genetic disorders.
Supplier: Charles River Laboratories Cell Solutions
Description: Whole blood is human blood from a standard blood donation. Whole blood is collected from IRB consented healthy human donors in FDA-registered collecti...
Catalog Number: (ROCKR500-0100)
Supplier: Rockland Immunochemicals
Description: Bovine/calf blood in heparin sodium.
UOM: 1 * 100 mL
Catalog Number: (ROCKR302-0050)
Supplier: Rockland Immunochemicals
Description: Chicken whole blood aseptically collected. These cell suspensions are ideal for washed pooled cells preparations, complement titration, adsorption pro...
UOM: 1 * 50 mL
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 4 of 4
no targeter for Bottom