You Searched For: Specimens
8  results were found
SearchResultCount:"8"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (ALEXBML-SW110-0200)
Supplier: Enzo Life Sciences
Description: Whole thymus extract from mouse designed for Western blotting control of anti-PARP antibodies or other antibodies/proteins.
UOM: 1 * 200 µl
Catalog Number: (ALEXADILYTHM100F)
Supplier: Enzo Life Sciences
Description: Microsomes are a rich source of heme-containing metabolic enzymes, including heme-oxygenases and cytochrome P450 (CYP) enzymes, and are commonly used ...
UOM: 1 * 200 µG
Catalog Number: (ALEXADILYTRB100F)
Supplier: Enzo Life Sciences
Description: Rat brain tissue extract
UOM: 1 * 200 µG
Supplier: Thermo Scientific
Description: Sapphire spheres, Grade: 25, Ø: 1,50 mm
Supplier: Thermo Scientific
Description: A single crystal is a material in which the crystal lattice of the entire sample is continuous and unbroken to the edges of the sample, with no grain ...
Catalog Number: (ALEXBML-SW115-0100)
Supplier: Enzo Life Sciences
Description: Whole cell extract of human Jurkat macrophage cells.
UOM: 1 * 100 µl
Catalog Number: (ALEXBML-SW106-0200)
Supplier: Enzo Life Sciences
Description: Whole liver extract from mouse useful as a Western blotting control for Aryl-hydrocarbon receptor antibodies or for use with other antibodies/proteins...
UOM: 1 * 200 µl
Catalog Number: (ICNA092850145)
Supplier: MP Biomedicals
Description: Chicken embryo extract prepared from nine to 12 day old eggs.
UOM: 1 * 20 mL
MSDS
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8 of 8
no targeter for Bottom