You Searched For: Autoclaves (Steam Sterilisers)
30  results were found
SearchResultCount:"30"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Autoclave for occasional use. AISI 304 stainless steel housing, easy to maintain and disinfect, aluminium inner chamber and ABS insulating lid with lo...
Sale
Supplier: VWR Collection
Description: VAPOUR-Line autoclaves are designed for applications such as liquids in open or slightly closed vessels, instruments and solids, unwrapped and waste s...
Supplier: VWR Collection
Description: Easy to use, automatic, flexible, reliable and convenient autoclave for all standard laboratory procedures with special features, such as Fâ‚€ calculati...
Supplier: VWR Collection
Description: These autoclaves are ideal for media, equipment and waste sterilisation. Designed for safety level laboratories, they are equipped with exhaust air fi...
Sale
Supplier: TUTTNAUER
Description: The Classic is reliable and trusted worldwide, designed to give you the most cost effective sterilisation solution. This light, compact, portable and ...
Supplier: TUTTNAUER
Description: Tuttnauer's laboratory autoclaves are ideal for a wide range of applications in various fields, such as research institutes, universities, pharmaceuti...
Supplier: TUTTNAUER
Description: Affordable performance and trusted reliability for your laboratory sterilisation needs. These compact units sterilise liquids, solids, sealed and unse...
Supplier: TUTTNAUER
Description: Tuttnauer's ML laboratory line of semi-automatic, electro-mechanically controlled, vertical steam sterilizers provide safe, economical and effective s...
Supplier: TUTTNAUER
Description: Tuttnauer's laboratory autoclaves are ideal for a wide range of applications in various fields, such as research institutes, universities, pharmaceuti...
Catalog Number: (TUTT3850EAP-D)
Supplier: TUTTNAUER
Description: The EA autoclave significantly increase your productivity with extra fast and efficient drying cycles. These two models have the added benefit of a hi...
UOM: 1 * 1 items
Catalog Number: (481-2004)
Supplier: TUTTNAUER
Description: Horizontal and vertical autoclave with automatic control. The chamber is constructed of long lasting 316L/Ti grade stainless steel with superior corro...
UOM: 1 * 1 items
Supplier: Priorclave
Description: These versatile autoclaves are fully equipped for use in laboratories throughout the food, drink, dairy, health and pharmaceutical industries and offe...
Supplier: Priorclave
Description: These autoclaves combine a huge internal capacity with a small floor area and are ideally suited for use in a wide range of sterilising applications s...
Supplier: SHP STERILTECHNIK AG
Description: A completely new concept allows the installation of the unit as either front or top loader. Deciding on horizontal or vertical is no longer the questi...
Supplier: SHP STERILTECHNIK AG
Description: These compact benchtop units with a 25 litre useable chamber offers a large capacity and the performance of a big autoclave. The unit has a chamber do...
Supplier: Priorclave
Description: Compact 40 benchtop and Compact 60 floor standing or benchtop autoclaves for the low volume user, with flexible operation usually only available in la...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 30
no targeter for Bottom