Microplates, glass coated, 96-well, WebSeal Plate+
548-3383EA 528 EUR
VWRI548-3383 VWRI548-3382 VWRI548-3380
Microplates, glass coated, 96-well, WebSeal Plate+
High quality polypropylene microplates coated with 200 nm thick layer of silicone dioxide.
  • Plate+ provides microplates with a chemical resistance similar to glass while retaining the advantages of polypropylene
  • Eliminates the need to use glass limited volume inserts
  • Excellent for applications where plastic microplates are not applicable
  • Plate+ is chemically and thermally resistant and will tolerate temperatures from −80 °C to +80 °C
  • Lightweight, precision moulded, cost effective alternative to solid glass plates
Other WebSeal products are available. Please contact your local VWR Office for further details.