Microinjectors, Recording Nanoject II™
DRUM3-000-205-RNEA 9560 EUR
DRUM3-000-205-RN
Microinjectors, Recording Nanoject II™
The Recording Nanoject II™ microinjector was specifically developed to improve micro-volume injection techniques used to study connectivity and function of specific brain nuclei. Designed to be operated in conjunction with an extracellular amplifier, the integrated system can be used to reliably place small volumes into targeted areas. It is engineered to easily mount on popular stereotaxic instruments.
  • Enables high precision
  • Facilitates connection to an extracellular amplifier to allow recording of neural activity
  • Supplements stereotaxic coordinates in guiding the placement of small injections
  • Provides direct displacement of neuronal tracer or excitotoxin from the pipette
  • Can be used on stereotaxic equipment utilising 7,9 mm clamps