Soldering tip, Plato® EW-402
Supplier: ITW Chemtronics
TEXWEW-402EA 0 EUR
TEXWEW-402
Soldering tip, Plato® EW-402
Soldering Supplies Soldering Tips
Manufactured from high grade copper stock machined to precise size. First, the tips are iron plated to resist solder erosion, then nickel plated for maximum corrosion protection. Chromium plating is then applied to further immunize the unwetted surfaces of the tip. Finally, the tip is tinned to provide maximum wetting and efficient heat transfer.
  • Interchangeable with Weller ET-series tips
  • For use with Weller WES50, WES51, WD1, WD2 soldering stations with EC1201A, EC1204A, PES50, PES51, WCC101 soldering handpieces
  • RoHS compliant