Disinfectant wipes, kodan® wipes
SCUE140401EA 9 EUR
SCUE140401
Disinfectant wipes, kodan® wipes
These ready to use wipes can be used for the disinfection and cleaning of skin and hands of patients, carers and visitors as well as for the disinfection of non medical areas such as the toilet area, on floors, door handles and other work surfaces. For use on larger areas, several wipes may be required.
  • Wipes can be removed individually and are ready to use immediately
  • Always rub the surface evenly and thoroughly with the wet wipe and then allow to work
  • Very broad antimicrobial effectiveness
  • Very good skin compatibility
  • Fast drying on surfaces
  • VAH/DGHM-listed
Microbiological effectiveness: bacteria incl. TB, fungi (Candida albicans), limited virucidal action (incl. HIV, HBV, HCV, herpes simplex viruses), influenza viruses (incl. influenza A/H1N1 and A/H5N1), rotaviruses, adenoviruses
100 g of active substance solution contains:
30,0 g of propan-2-ol, 30,0 g of propan-1-ol
Please note: Handle disinfectants with care. Always read the label and product information before use. Should not be used on alcohol sensitive surfaces.
Caution: Use biocides safely. Always read the label and product information before use.