Cleanroom anti-fatigue mats, Ergomat® Plano Conductive
Supplier: Ergomat
Ergomat®
ERGOPLANOCON-BLACKEA 155 EUR
ERGOPLANOCON-BLACK
Cleanroom anti-fatigue mats, Ergomat® Plano Conductive
Floor Mats Traction and Anti-Fatigue Floor Mats
The Ergomat® Plano Conductive is a smooth mat made from pure nitrile rubber and is designed to increase comfort while reducing tripping hazards, body aches, quality issues, and sick leaves. It is ideal for usage in the electronics industry or for areas where static control is crucial due to sensitive equipment. Ideal for areas where trolleys and carts are used.
  • Great anti-bacterial cleaning properties
  • Chemical exposure will not affect the state of the mat; no curling up, swelling, or expanding
  • Moulded bevelled edges prevent curling, eliminates trip hazards
  • Raw virgin material guarantee durability and ergonomic benefits
  • Free from silicone and latex
Suitable for use in Class ISO 5 (100) cleanrooms.
Colour: Black
Certifications: Meets EN ISO/USDA/CFIA standards