Light boxes, Prolite Basic 2
KAFO2425EA 680 EUR
KAFO2425 KAFO2403
Light boxes, Prolite Basic 2
The prolite basic two light boxes provide brilliant, homogenous and professional illumination.
  • All-metal housing is rigid and durable and also wall mountable
  • Totally flat surface
  • Lamps 5400 K, colour rendition group 1A (CRI range 90 to 100)
  • AC operation