Temperature and humidity loggers, compact
Supplier: Dickson
DICKTK550EA 1430 EUR
DICKTK550
Temperature and humidity loggers, compact
Data Loggers
This 12-pack of coin-sized temperature and humidity data loggers provide a great per unit price making them ideal for mapping warehouses or large stability chambers. Thanks to their small size they are ideal for shipping and transportation applications.
  • EEPROM memory type
  • Easy to use
  • Two channels: Temperature and humidity
  • Polycarbonate enclosure
  • Download time of 30 s
Certifications: IP 20 certified protection class
Ordering information: Supplied with three volt lithium battery (one per unit) and Quick Start Guide. Software is needed for operation but not included. It can be ordered separately. PC requirements: Microsoft Windows 98 or newer; 16 MB RAM.