Universal safety heating bath with integrated carrying handles and safety temperature circuit, mainly used for water and oil applications.
- Ergonomically designed handles
- Safety temperature cut-off
- Lock function prevents accidental change of set conditions
- Micro-controller temperature control
- Optimised bath volume (3 L) guarantees fast heat-up
- Accuracy: ±1 K
The built-in dry run control stops the bath when there is no medium present. Laterally arranged carrying handles allow for safe handling without the risk of burns. The heating bath is designed with high-quality, recyclable materials and therefore contributes to resource-preserving production.