ESD finger cots, nitrile, QRP®
Supplier: QRP
112-4192EA 106 EUR
QUAL9CS QUAL9CM QUAL9CXL 112-4192
ESD finger cots, nitrile, QRP®
Finger Cots Nitrile Finger Cots
These latex-free finger cots are packaged for ISO 5 (Class 100) cleanroom applications. They are ideal for use with Class II static-sensitive devices (thresholds above 1000 V). Suitable for use in IC industries, wire bonding SMT, WB assembly and for discrete component handling.
  • Powder-free
  • Average surface resistance of 5×10⁹ ohms per square unit
  • Excellent splash and intermittent protection from many common chemicals and solvents like IPA and acetone
  • Consistently low extractables and ionics
Colour: Natural