iBONi® siRNA controls
Supplier: riboxx Life Sciences
XXXXK-00401-0002-H
iBONi® siRNA controls
iBONi® siRNA controls are designed to be used as positive, negative or transfection controls for validation of gene silencing experiments. iBONi® siRNAs controls originate either from published experimental data or have been successfully tested by independent scientists in human, mouse and rat cells. All iBONi® siRNA controls include the riboxx® siRNA design, which increases the potency of the siRNAs and reduces off-target effects.