Salt tester, SALINTEST, HI 98203
Supplier: Hanna
HANNHI98203EA 200 EUR
HANNHI98203
Salt tester, SALINTEST, HI 98203
Electrochemical Meters Salinity Meters
This meter accurately monitors the salinity, the concentration of sodium chloride, in aquaculture systems.
  • Easy to maintain
  • One point calibration through a trimmer on the side
  • Replaceable electrode
Besides applications in aquaculture, it is also ideal for checking salt concentrations in live fish storage tanks, tropical fish aquariums, refrigerated storage and oceanographic investigations.
Delivery information: Supplied with protective cap, calibration screwdriver, batteries, instructions and with a handy chart that converts readings into g/L of sodium chloride.