Ovens, MINO, Digital series
Supplier: Genlab
466-0442EA 2220 EUR
466-0439 466-0442 466-0441 466-0437 466-0438 466-0440 466-0436
Ovens, MINO, Digital series
Ovens Standard Ovens
Reliable, efficient, general purpose ovens, to suit most drying, warming and general laboratory applications. These units from Genlab have a sheet steel exterior finished with an easy to clean powder coated paint. The interior chamber is made from mild steel coated with aluminium (CLAD). They are heated via Incoloy sheathed elements fitted around the fan on the rear of the chamber. Ovens have fixed shelf runners and removable chrome plated wire grid shelves. The top vent is fitted with a clip to hold a glass immersion thermometer.
  • Temperature fluctuation ±0,5 °C
  • Temperature range 40 to 250 °C
  • Microprocessor digital controller with dual display of set point and actual temperature
  • Independent adjustable temperature safety system with visual alarm protects the samples and the oven
The units are fitted with a PID microprocessor controller with dual displays of set point and actual temperature, which are auto tuned for each individual unit to optimise the heat up, overshoot and control of temperature. They also include main switch with indicator and heat and overheat indicators.
Delivery information: Supplied with removable chrome plated wire grid shelves. Options such as a stainless steel interior and process timer are available, please contact your local VWR office for details.