Electromantles for heating and stirring with the added functionality of magnetic stirring. For convenience and safety, the electric circuitry for stirring is built into the base of the heater, enabling powerful stirring over a wide range of solution volumes and viscosities using a magnetic stir bar.
- Stir speed is bi-directional and easily adjustable with auto-recapture switch
- Stirring speed up to 2000 min⁻¹
- Element temperatures up to 450 °C
Built-in energy regulator with indicator lamps for power and heater operation. An easy to clean powder coated aluminium outer casing and a non-skid feet and support rod bracket. Added safety features such as a grounded earth screen and double fuses.