You Searched For: BOD Incubators
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: WTW
Description: These thermostatic cabinets are available in three sizes and different variants. Suited for BOD, OECD 301F, soil respiration and anaerobic degradatio...
Supplier: Lovibond
Description: The TC series of thermostatically controlled cabinets is used for continuous temperature control over a range of 2 to 40 °C. This makes them ideal for...
Catalog Number: (VELPF10300305)
Supplier: VELP SCIENTIFIC
Description: These BOD incubators have been designed to keep any product at a constant temperature, where accurate and precise temperature control is required. For...
UOM: 1 * 1 items
Catalog Number: (WITGDH.WIR03250)
Supplier: WITEG LABORTECHNIK GMBH
Description: Low temperature BOD incubator with superior temperature uniformity by forced convection.
UOM: 1 * 1 items
Catalog Number: (VWRU35960-056)
Supplier: VWR Collection
Description: These refrigerated incubators with microprocessor control are widely used for applications such as BOD determinations, plant and insect studies, ferme...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom