ESD occupational shoes, lace-up, Light, 31151
Supplier: Abeba
ABEB31151/36EA 95.4 EUR
ABEB31151/36
ESD occupational shoes, lace-up, Light, 31151
ESD shoe made from smooth leather and a lightweight, fuel resistant PU sole.
  • Temperature regulating special lining Comfortemp®
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbent heel
Colour: White
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; O2, FO, SRA