Tubing, LDPE
Supplier: Semadeni
SEMA2382EA 101 EUR
SEMA2382 228-2398
Tubing, LDPE
Tubing Transfer Tubing
Made of LDPE, translucent
  • Slightly flexible
  • Highly resistant to chemicals and solvents
  • Good temperature resistance