VWR® HistoChoice®, Mounting media
Supplier: Avantor
HistoChoice®
H157-475MLEA 72.9 EUR
H157-475ML
VWR® HistoChoice®, Mounting media
Stains Microscopy and staining reagents
HistoChoice® mounting media was designed for use with HistoChoice® clearing agent, but can also be used as a separate, stand-alone medium for standard slide preparation. It is suitable for both thick and thin specimen mounting. It can also be used with immersion oil.
  • Low viscosity means easier application resulting in bubble-free preparations which saves valuable laboratory time