High purity nitrogen generators
Supplier: Parker
Parker domnick hunter
564-0132EA 13710 EUR
564-0132 564-0137 564-0136 564-0139 564-0138
High purity nitrogen generators
Gas Generators Nitrogen Generators
These nitrogen gas generators employ robust, field proven technology to produce high purity nitrogen for life science, chemical analysis and spectroscopy applications. Flow rates range from 5 to 12,5 L/min, with purities from 99,5 to 99,999%. The generators provide a continuous stream of high purity nitrogen from a single 'plug & play' unit. Models are available with and without an integral oil free compressor.
  • Economy mode as standard, increases compressor life
  • Integral oil-free compressor, with advanced noise reduction technology
  • Eliminate inconvenient and potentially dangerous nitrogen cylinders
Suffix ‘E' for 207 to 253 V 50/60 Hz
Suffix 'W' for 103 to 126 V 60 Hz.
Suffix"‘E" for 207 - 253 V 50/60 Hz
Suffix "W" for 103 - 126 V 60 Hz