Ultra high purity zero nitrogen generators
Supplier: Parker
Parker domnick hunter
564-0166EA 12281 EUR
564-0166 564-0164 564-0163 564-0162 564-0161 564-0160
Ultra high purity zero nitrogen generators
Gas Generators Nitrogen Generators
These generators are designed for GC makeup gas and carrier gas applications. An integral heated platinum catalyst ensures carrier grade nitrogen free from organic impurities to <0,1 ppm. The generators provide a continuous stream of ultra high purity nitrogen from a single 'plug & play' unit. Models are available with and without an integral oil free compressor.
  • Ultra high purity (>99,999%), organic-free, nitrogen
  • Economy mode as standard, increases compressor life
  • Integral oil-free compressor, with advanced noise reduction technology
  • Eliminate inconvenient and potentially dangerous nitrogen cylinders
Suffix 'E' for 207 to 253 V 50/60 Hz
Suffix 'W' for 103 to 126 V 60 Hz