Finger cots, latex, LWR
Supplier: North by Honeywell
112-3908EA 53.5 EUR
112-3908
Finger cots, latex, LWR
Finger Cots Latex Finger Cots
These natural rubber finger cots are suitable for many different applications. The thin gauge makes them perfect for extremely sensitive tasks, such as battery manufacturing, electrical work, food processing, laboratory work and photo processing.
  • Powdered
  • Thin gauge for extremely high sensitivity
  • Tapered tip is more comfortable and provides better fit
  • In accordance with FDA regulations for food contact
Thickness: 0,076 mm
Length: 70 mm
Thickness: 0,076 mm
Colour: White