Occupational shoes, clogs, X-Light, 711141/711142
Supplier: Abeba
X-Light
ABEB711142/40EA 75.9 EUR
ABEB711142/40 ABEB711142/42 ABEB711142/41 ABEB711142/48 ABEB711142/37 ABEB711142/36 ABEB711142/47 ABEB711142/39 ABEB711141/38 ABEB711142/38 ABEB711142/44 ABEB711142/43 ABEB711142/46 ABEB711142/35 ABEB711142/45
Occupational shoes, clogs, X-Light, 711141/711142
Shoes and Boots
Anti-static shoes made from smooth leather upper and and a lightweight, fuel resistant PU sole.
  • Perspiration-absorbent lining with silver point
  • Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbing system
  • Foldable and adjustable heel strap
Colour: White or black
Certifications: DGUV 112-191; EN ISO 20347:2012; OB, A, E, FO, SRC