Infrared turbidity meter, TurbiCheck
Supplier: Lovibond Tintometer
LOVI266020EA 602 EUR
LOVI266020
Infrared turbidity meter, TurbiCheck
TurbiCheck is designed for fast and precise on-site testing.
  • For measurements of coloured liquids
  • 600 tests without battery change
  • Integrated real time and calendar
Certifications: Infrared light with an angle of 90° as per ISO 7027.
Delivery information: Supplied with four turbidity standards (0,1; 20; 200; 800 NTU), battery and test vials in a case.