Certified thermometer
Supplier: Amarell
AMARA307310EA 0 EUR
AMARA307310
Certified thermometer
This glass thermometer is an alternative to the government verified instruments.
This glass thermometer is an alternative to the government verified instruments.
Filling: Mercury
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.